missing translation for 'onlineSavingsMsg'
Learn More

6-Mercaptopurine hydrate, MedChemExpress

Product Code. 16481686
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481686 500 mg 500mg
16461957 50 mg 50mg
16436276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481686 Supplier Medchem Express Supplier No. HY13677A
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 6-Mercaptopurine hydrate (Mercaptopurine hydrate; 6-MP hydrate) is a purine analogue which acts as an antagonist of the endogenous purines and has been widely used as antileukemic agent and immunosuppressive drug.

6-Mercaptopurine hydrate (Mercaptopurine hydrate

Specifications

Chemical Name or Material 6-Mercaptopurine hydrate
Physical Form Solid
CAS 6112-76-1
Color Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C5H6N4OS
For Use With (Application) Cancer-programmed cell death
Synonym Mercaptopurine hydrate 6-MP hydrate
Solubility Information DMSO : 50 mg/mL (293.79 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES S=C1NC=NC2=C1NC=N2.O
Molecular Weight (g/mol) 170.19
Formula Weight 170.19
Percent Purity 98.61%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.