missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-tetralone, 90%

Product Code. 10752961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10752961 1 g Glass bottle 1g
10025993 250 mg Glass bottle 250mg
10219253 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10752961 Supplier Thermo Scientific Acros Supplier No. 298920010
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2472-22-2 | C11H12O2 | 176.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2472-22-2
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00001729
InChI Key RMRKDYNVZWKAFP-UHFFFAOYSA-N
Synonym 6-methoxy-2-tetralone, 6-methoxy-3,4-dihydronaphthalen-2 1h-one, 6-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 6-methoxyl-2-tetralone, 3,4-dihydro-6-methoxy-2 1h-naphthalenone, 6-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-6-methoxynaphthalen-2 1h-one, 6-methoxy-1,3,4-trihydronaphthalen-2-one
PubChem CID 75582
IUPAC Name 6-methoxy-3,4-dihydro-1H-naphthalen-2-one
SMILES COC1=CC2=C(CC(=O)CC2)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 28°C to 35°C
Color Beige-Yellow to Orange
Boiling Point 114°C to 116°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 176.22
Percent Purity 90%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 6-Methoxy-2-tetralone
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methoxy-2-tetralone
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/physician/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.