missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-tetralone, tech. 90%, Thermo Scientific™

Product Code. 11363136
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363136 250 mg 250mg
11353136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363136 Supplier Thermo Scientific Alfa Aesar Supplier No. L11007.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2472-22-2
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00001729
InChI Key RMRKDYNVZWKAFP-UHFFFAOYSA-N
Synonym 6-methoxy-2-tetralone, 6-methoxy-3,4-dihydronaphthalen-2 1h-one, 6-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 6-methoxyl-2-tetralone, 3,4-dihydro-6-methoxy-2 1h-naphthalenone, 6-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-6-methoxynaphthalen-2 1h-one, 6-methoxy-1,3,4-trihydronaphthalen-2-one
PubChem CID 75582
IUPAC Name 6-methoxy-3,4-dihydro-1H-naphthalen-2-one
SMILES COC1=CC2=C(CC(=O)CC2)C=C1

Specifications

Melting Point ∼35°C
Boiling Point 114°C to 116°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.561
Quantity 250 mg
Beilstein 513418
Formula Weight 176.22
Percent Purity 90%
Grade Technical
Chemical Name or Material 6-Methoxy-2-tetralone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.