missing translation for 'onlineSavingsMsg'
Learn More

6-Methylindole, 99%

Product Code. 10366540 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366540 1 g Glass bottle 1g
10061340 250 mg Glass bottle 250mg
10075993 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10366540 Supplier Thermo Scientific Acros Supplier No. 133880010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3420-02-8 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3420-02-8
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00005682
InChI Key ONYNOPPOVKYGRS-UHFFFAOYSA-N
Synonym n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester
PubChem CID 137930
SMILES CC1=CC=C2C=CNC2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.0500g/mL
Boiling Point 112°C (5.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6060 to 1.608
Quantity 1 g
Beilstein 20, IV, 3214
Specific Gravity 1.05
IUPAC Name tert-butyl N-phenylcarbamate
Formula Weight 131.18
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 6-Methylindole
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methylindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.