missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpyridine-3-boronic Acid Hydrochloride Hydrate, TRC

Product Code. 30465116
Change view
Click to view available options
Quantity:
10 mg
1 g
5 g
Unit Size:
10mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465116 1 g 1g
30466955 5 g 5g
30470934 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30465116 Supplier Toronto Research Chemicals Supplier No. TRCM3270901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methylpyridine-3-boronic Acid Hydrochloride Hydrate
Analyte or Component Names 6-Methylpyridine-3-boronic Acid Hydrochloride Hydrate
Shipping Condition Room Temperature
Molecular Formula C6H8BNO2. HCl . xH 2O
Synonym B-(2-Methylpyridin-5-yl)boronic Acid Hydrochloride Hydrate, (2-Methylpyridin-5-yl)boronic Acid Hydrochloride Hydrate, (6-Methyl-3-pyridinyl)boronic Acid Hydrochloride Hydrate, 2-Methyl-5-pyridinylboronic Acid Hydrochloride Hydrate, 6-Methyl-3-pyridineboronic Acid Hydrochloride Hydrate
InChI Formula InChI=1S/C6H8BNO2.ClH.H2O/c1-5-2-3-6(4-8-5)7(9)10;;/h2-4,9-10H,1H3;1H;1H2
Solution Type Neat
SMILES OB(C1=CC=C(N=C1)C)O.Cl.O
IUPAC Name (6-methylpyridin-3-yl)boronic acid hydrochloride hydrate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 191.42
Quantity 1 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.