missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Hepatitis B Antigen Protein

Product Code. 18696237 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18696237

Brand: Novus Biologicals™ NBP262489

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) Western Blot, ELISA
Formulation PBS
Molecular Weight (g/mol) MolecularWeight-theroretical: 24 kDa
Quantity 0.1 mg
Storage Requirements Store at 4°C.
Gene Alias HBV x antigen, HBx, HBxAg
Purity or Quality Grade >95%, by SDS-PAGE
Protein Hepatitis B Antigen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.