missing translation for 'onlineSavingsMsg'
Learn More

6-(tert-Butyl) 3-ethyl 2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate, Thermo Scientific™

Product Code. 10092124
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10092124 5 g Amber Glass Bottle 5g
10423763 250 mg Amber Glass Bottle 250mg
10433763 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092124 Supplier Thermo Scientific Maybridge Supplier No. MO01303DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 193537-14-3
Molecular Formula C15H22N2O4S
Molecular Weight (g/mol) 326.411
InChI Key XSXVOVXVHBSSSN-UHFFFAOYSA-N
Synonym ethyl 6-boc-2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3-carboxylate, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4,5-dihydrothieno 2,3-c pyridine-3,6 7h-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4h,5h,6h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4,7-dihydrothieno 2,3-c pyridine-3,6 5h-dicarboxylate, 2-amino-6-boc-4,5,6,7-tetrahydro-thieno 2,3-c pyridine-3-carboxylic acid ethyl ester, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert-butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4h,5h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, ethyl 2-amino-4,5,6,7-tetrahydrothieno 2,3-c pyridine-3-carboxylate, n-boc protected
PubChem CID 2794740
IUPAC Name 6-O-tert-butyl 3-O-ethyl 2-amino-5,7-dihydro-4H-thieno[2,3-c]pyridine-3,6-dicarboxylate
SMILES CCOC(=O)C1=C(SC2=C1CCN(C2)C(=O)OC(C)(C)C)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 326.41
Percent Purity 95%
Chemical Name or Material 6-(tert-Butyl) 3-ethyl 2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.