missing translation for 'onlineSavingsMsg'
Learn More

6-(tert-Butylamino)-3-methyl-1,3,5-triazine-2,4(1H,3H)-dione, TRC

Product Code. 30135233
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135233 50 mg 50mg
30009406 5 mg 5mg
30004399 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135233 Supplier Toronto Research Chemicals Supplier No. TRCB18596050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206682-85-9
Molecular Formula C8 H14 N4 O2
Molecular Weight (g/mol) 198.222
Synonym Terbuthylazine Metabolite SYN 545666, 1,3,5-Triazine-2,4(1H,3H)-dione, 6-[(1,1-dimethylethyl)amino]-3-methyl-, Terbuthylazine Metabolite LM6
IUPAC Name 4-(tert-butylamino)-6-hydroxy-1-methyl-1,3,5-triazin-2-one
SMILES CN1C(=NC(=NC1=O)NC(C)(C)C)O

Specifications

Analyte or Component Names Terbuthylazine Metabolite SYN 545666
Shipping Condition Room Temperature
InChI Formula InChI=1S/C8H14N4O2/c1-8(2,3)11-5-9-6(13)12(4)7(14)10-5/h1-4H3,(H2,9,10,11,13,14)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Formula Weight 198.1117
Percent Purity >95%
Chemical Name or Material Terbuthylazine Metabolite SYN 545666
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.