missing translation for 'onlineSavingsMsg'
Learn More

6alpha,9alpha-Difluoroprednisolone 21-Acetate, TRC

Product Code. 30370135
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370135 250 mg
30369081 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370135 Supplier Toronto Research Chemicals Supplier No. TRCD446325250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-70-0
Molecular Formula C23H28F2O6
Molecular Weight (g/mol) 438.46
Synonym (6α,11β)-21-(Acetyloxy)-6,9-difluoro-11,17-dihydroxypregna-1,4-diene-3,20-dione, 6α,9-Difluoro-11β,17,21-trihydroxypregna-1,4-diene-3,20-dione
SMILES O=C(COC(C)=O)[C@@]1(O)CC[C@@]2([H])[C@]3([H])C[C@H](F)C4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@@]21C)=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C23H28F2O6/c1-12(26)31-11-19(29)22(30)7-5-14-15-9-17(24)16-8-13(27)4-6-20(16,2)23(15,25)18(28)10-21(14,22)3/h4,6,8,14-15,17-18,28,30H,5,7,9-11H2,1-3H3/t14-,15-,17-,18-,20-,21-,22-,23-/m0/s1
Formula Weight 438.1854
Chemical Name or Material 6alpha,9alpha-Difluoroprednisolone 21-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.