missing translation for 'onlineSavingsMsg'
Learn More

6Alpha-Methyl Prednisolone 17-Methylhemisuccinate, TRC

Product Code. 30475594
Change view
Click to view available options
Quantity:
5 mg
25 mg
250 mg
Unit Size:
250mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475594 25 mg 25mg
30476126 5 mg 5mg
30484049 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475594 Supplier Toronto Research Chemicals Supplier No. TRCM30385525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6alpha-Methyl Prednisolone 17-Methylhemisuccinate
Analyte or Component Names 6alpha-Methyl Prednisolone 17-Methylhemisuccinate
Shipping Condition Room Temperature
Molecular Formula C27H36O8
InChI Formula InChI=1S/C27H36O8/c1-15-11-17-18-8-10-27(21(31)14-28,35-23(33)6-5-22(32)34-4)26(18,3)13-20(30)24(17)25(2)9-7-16(29)12-19(15)25/h7,9,12,15,17-18,20,24,28,30H,5-6,8,10-11,13-14H2,1-4H3/t15-,17-,18-,20-,24+,25-,26-,27-/m0/s1
Solution Type Neat
SMILES [H][C@@]12[C@]([C@](C=C3)(C)C([C@@H](C)C2)=CC3=O)([H])[C@@H](O)C[C@@]4(C)[C@@]1([H])CC[C@@]4(C(CO)=O)OC(CCC(OC)=O)=O
IUPAC Name (6S,8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl methyl succinate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 488.57
Quantity 25 mg
Formula Weight 488.24
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.