missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-1,3-naphthalenedisulfonic acid, Tech.

Product Code. p-58584 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10696172

Brand: Thermo Scientific Acros 163961000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-65-7 | C10H9NO6S2 | 303.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-65-7
Molecular Formula C10H9NO6S2
Molecular Weight (g/mol) 303.29
MDL Number MFCD00003991
InChI Key CMOLPZZVECHXKN-UHFFFAOYSA-N
Synonym amido-g-acid, 7-amino-1,3-naphthalenedisulfonic acid, amino-g-acid, 1,3-naphthalenedisulfonic acid, 7-amino, amino-g acid, 2-amino-6,8-disulfonaphthalene, unii-7c482fx29k, 2-aminonaphthalene-6,8-disulfonic acid, 2-naphthylamine-6,8-disulfonic acid, beta-naphthylamine-6,8-disulfonic acid
PubChem CID 6851
IUPAC Name 7-aminonaphthalene-1,3-disulfonic acid
SMILES C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)S(=O)(=O)O)N

Specifications

Melting Point >300°C
Color Brown to White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Beilstein 14, 784
Merck Index 15, 395
Solubility Information Solubility in water: soluble
Formula Weight 303.29
Percent Purity 60% (as is basis) Typically
Grade Technical
Physical Form Needles or Powder
Chemical Name or Material 7-Amino-1, 3-naphthalenedisulfonic acid, tech.
Show More Show Less
compliance-icons
Product Identifier
  • 7-Amino-1,3-naphthalenedisulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.