missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-4-methylcoumarin, Laser Grade 99%, ACROS Organics™

Product Code. 10025790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10025790 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10025790 Supplier Thermo Scientific Acros Supplier No. 405555000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 26093-31-2
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C10H9NO2
MDL Number MFCD00006868
Quantity 500 mg
Beilstein 18, V,11, 445
Synonym 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x
InChI Key GLNDAGDHSLMOKX-UHFFFAOYSA-N
SMILES CC1=CC(=O)OC2=CC(N)=CC=C12
IUPAC Name 7-amino-4-methyl-2H-chromen-2-one
Molecular Weight (g/mol) 175.19
PubChem CID 92249
ChEBI CHEBI:51771
Formula Weight 175.19
Percent Purity ≥98.5%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material 7-Amino-4-methylcoumarin, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.