missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-3,4-dihydronaphthalen-1(2H)-one, 97%, Thermo Scientific Chemicals

Product Code. 13525030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13525030 50 g Glass bottle 50g
13505030 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13525030 Supplier Thermo Scientific Acros Supplier No. 448680500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32281-97-3
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.08
InChI Key YGVDCGFUUUJCDF-UHFFFAOYSA-N
Synonym 7-bromo-1-tetralone, 7-bromo-3,4-dihydronaphthalen-1 2h-one, 7-bromo-3,4-dihydro-1 2h-naphthalenone, 7-bromotetralone, 7-bromo-alpha-tetralone, 7-bromotetralin-1-one, 7-bromo-1,2,3,4-tetrahydronaphthalen-1-one, 7-bromo-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone, 7-bromo-3,4-dihydro, 7-bromo tetralone
PubChem CID 252731
IUPAC Name 7-bromo-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=C(C=C2)Br)C(=O)C1

Specifications

Melting Point 78.0°C to 79.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Formula Weight 225.08
Percent Purity 97%
Chemical Name or Material 7-Bromo-3, 4-dihydronaphthalen-1(2H)-one
compliance-icons
Product Identifier
  • 7-Bromo-3,4-dihydronaphthalen-1(2H)-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.