missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-quinoline-3-carboxylic Acid, TRC

Product Code. 30133787
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133787 10 g 10g
30136710 25 g 25g
17912965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133787 Supplier Toronto Research Chemicals Supplier No. TRCC36513510G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86393-33-1
Molecular Formula C13 H9 Cl F N O3
Molecular Weight (g/mol) 281.67
Synonym 7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Enrofloxacin Imp. A (EP), Ciprofloxacin Hydrochloride Imp. A (EP), Fluoroquinolonic Acid
IUPAC Name 7-chloro-1-cyclopropyl-6-fluoro-4-oxoquinoline-3-carboxylic acid
SMILES OC(=O)C1=CN(C2CC2)c3cc(Cl)c(F)cc3C1=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H9ClFNO3/c14-9-4-11-7(3-10(9)15)12(17)8(13(18)19)5-16(11)6-1-2-6/h3-6H,1-2H2,(H,18,19)
Formula Weight 281.0255
Percent Purity >95%
Chemical Name or Material Fluoroquinolonic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.