accessibility menu, dialog, popup

UserName

OONClFO

CAS RN 86393-33-1

IUPAC Name: 7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
Synonyms: 7-chloro-1-cyclopropyl-6-fluoro-4-oxoquinoline-3-carboxylate
Molecular Weight (g/mol): 280.66
Molecular Formula: C13H8ClFNO3
InChi Key: ISPVACVJFUIDPD-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CN(C2CC2)C2=CC(Cl)=C(F)C=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results