missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-3-iodo Hydroxychloroquine, TRC

Product Code. 30350346
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350346 50 mg 50mg
30357669 5 mg 5mg
30358332 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350346 Supplier Toronto Research Chemicals Supplier No. TRCC29346050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161618-30-0
Molecular Formula C18H25ClIN3O
Molecular Weight (g/mol) 461.77
Synonym 2-((4-((7-Chloro-3-iodoquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol
IUPAC Name 2-[4-[(7-chloro-3-iodoquinolin-4-yl)amino]pentyl-ethylamino]ethanol
SMILES CCN(CCO)CCCC(NC1=C2C=CC(Cl)=CC2=NC=C1I)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H25ClIN3O/c1-3-23(9-10-24)8-4-5-13(2)22-18-15-7-6-14(19)11-17(15)21-12-16(18)20/h6-7,11-13,24H,3-5,8-10H2,1-2H3,(H,21,22)
Chemical Name or Material 7-Chloro-3-iodo Hydroxychloroquine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.