missing translation for 'onlineSavingsMsg'
Learn More

7-Deoxy Doxorubicin Aglycone (>75%), TRC

Product Code. 30368234
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368234 1 mg 1mg
30359653 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30368234 Supplier Toronto Research Chemicals Supplier No. TRCD2329101MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38554-25-5
Molecular Formula C21 H18 O8
Molecular Weight (g/mol) 398.36
Synonym 5,12-Naphthacenedione, 7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-, (8R)-, 5,12-Naphthacenedione, 7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-(hydroxyacetyl)-1-methoxy-, (8R)- (9CI), 5,12-Naphthacenedione, 7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-(hydroxyacetyl)-1-methoxy-, (R)-, (8R)-7,8,9,10-Tetrahydro-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,12-naphthacenedione, 7-Deoxyadriamycin aglycone, 7-Deoxyadriamycinone, 7-Deoxydoxorubicin aglycone, 7-Deoxydoxorubicinone, 7-Deoxyepirubicin aglycone, NSC 270536
IUPAC Name (9R)-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione
SMILES COc1cccc2C(=O)c3c(O)c4C[C@](O)(CCc4c(O)c3C(=O)c12)C(=O)CO

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H18O8/c1-29-12-4-2-3-10-14(12)20(27)16-15(18(10)25)19(26)11-7-21(28,13(23)8-22)6-5-9(11)17(16)24/h2-4,22,24,26,28H,5-8H2,1H3/t21-/m1/s1
Formula Weight 398.1002
Percent Purity >95%
Chemical Name or Material 7-deoxy Doxorubicin Aglycone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.