missing translation for 'onlineSavingsMsg'
Learn More

7-Diethylamino-4-methylcoumarin, 99%

Product Code. 11435037
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435037 100 g 100g
11445037 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435037 Supplier Thermo Scientific Alfa Aesar Supplier No. B21938.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-44-1 | C14H17NO2 | 231.295 g/mol

7-(Diethylamino)-4-methylcoumarin, is potentially useful as a laser dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-(Diethylamino)-4-methylcoumarin, is potentially useful as a laser dye.

Solubility
Soluble in water, aqueous acid solutions, resins, varnishes, vinyls, ether, acetone, ordinary organic solvents, and alcohol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-44-1
Molecular Formula C14H17NO2
Molecular Weight (g/mol) 231.295
MDL Number MFCD00006864
InChI Key AFYCEAFSNDLKSX-UHFFFAOYSA-N
Synonym 7-diethylamino-4-methylcoumarin, coumarin 1, 7-diethylamino-4-methyl-2h-chromen-2-one, coumarin 47, coumarin 460, 4-methyl-7-diethylamino coumarin, blancophor aw, blancophor ffg, 4-methyl-7-diethylaminocoumarin, hakkol p
PubChem CID 7050
ChEBI CHEBI:51938
IUPAC Name 7-(diethylamino)-4-methylchromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C

Specifications

Melting Point 72°C to 75°C
Quantity 100 g
Beilstein 193303
Solubility Information Soluble in water,aqueous acid solutions,resins,varnishes,vinyls,ether,acetone,ordinary organic solvents,and alcohol.
Formula Weight 231.3
Percent Purity 99%
Chemical Name or Material 7-Diethylamino-4-methylcoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.