missing translation for 'onlineSavingsMsg'
Learn More

7-Ethoxycoumarin, 99%

Product Code. 11338985
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338985 100 mg 100mg
11328985 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338985 Supplier Thermo Scientific Alfa Aesar Supplier No. L14116.MC
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 31005-02-4 | C11H10O3 | 190.198 g/mol

7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.

Solubility
Soluble in hot methanol (very faint turbidity), 95% ethanol (50 mg/ml), DMF, and DMSO. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31005-02-4
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00006877
InChI Key LIFAQMGORKPVDH-UHFFFAOYSA-N
Synonym 7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx
PubChem CID 35703
ChEBI CHEBI:28184
IUPAC Name 7-ethoxychromen-2-one
SMILES CCOC1=CC2=C(C=C1)C=CC(=O)O2

Specifications

Melting Point 88°C to 91°C
Quantity 100 mg
Beilstein 146200
Solubility Information Soluble in hot methanol (very faint turbidity),95% ethanol (50mg/ml),DMF,and DMSO. Insoluble in water.
Formula Weight 190.2
Percent Purity 99%
Chemical Name or Material 7-Ethoxycoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.