missing translation for 'onlineSavingsMsg'
Learn More

7-Fluorooxindole, 95%

Product Code. 15498068
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498068 250 mg 250mg
15478068 1 g 1g
15488068 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498068 Supplier Thermo Scientific Alfa Aesar Supplier No. H61511.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 71294-03-6 | C8H6FNO | 151.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71294-03-6
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD02179608
InChI Key VMUIOEOYZHJLEZ-UHFFFAOYSA-N
Synonym 7-fluorooxindole, 7-fluoroindolin-2-one, 7-fluoro-oxindole, 7-fluoro-2-oxindole, 7-fluoroindolinone, 7-fluoro-1,3-dihydro-2h-indol-2-one, 2h-indol-2-one, 7-fluoro-1,3-dihydro, 7-fluoro-1,3-dihydro-indol-2-one, 7-fluoro-2,3-dihydro-1h-indol-2-one, 1,3-dihydro-7-fluoro-2h-indol-2-one
PubChem CID 3734371
IUPAC Name 7-fluoro-1,3-dihydroindol-2-one
SMILES C1C2=C(C(=CC=C2)F)NC1=O

Specifications

Color Red to Brown
Quantity 250 mg
Formula Weight 151.14
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 7-Fluorooxindole

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.