missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-3,4-dihydro-2(1H)-quinolinone, MedChemExpress

Product Code. 16429346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16429346 Supplier Medchem Express Supplier No. HYW010130

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 7-​Hydroxy-​3,​4-​dihydro-​2(1H)​-​quinolinone (3,4-Dihydro-7-hydroxy-2(1H)-quinolinone) is a weak MAO-A inhibitor, with an IC50 of 183 μM, and has no effect on MAO-B.

7-​Hydroxy-​3,​4-​dihydro-​2(1H)​-​quinolinone (3,4-Dihydro-7-hydroxy-2(1H)-quinolinone) is a weak MAO-A inhibitor, with an IC50 of 183 μM, and has no effect on MAO-B.

Specifications

Chemical Name or Material 7-Hydroxy-3,4-dihydro-2(1H)-quinolinone
CAS 22246-18-0
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C9H9NO2
Quantity 100 mg
Synonym 3,4-Dihydro-7-hydroxy-2(1H)-quinolinone
Solubility Information DMSO : 100 mg/mL (612.86 mM; Need ultrasonic) ∣H2O : 1 mg/mL (6.13 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=CC(O)=C2)CC1
Molecular Weight (g/mol) 163.17
Formula Weight 163.17
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.