missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-4'-nitroisoflavone, 98%, Thermo Scientific™

Product Code. 15431249
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431249 100 g 100g
15411249 5 g 5g
15421249 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431249 Supplier Thermo Scientific Alfa Aesar Supplier No. H64167.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
7-Hydroxy-4′-nitroisoflavone is used in the synthesis of iso flavones, Synthesis of daidzin, soflavone amide derivatives.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15485-80-0
Molecular Formula C15H9NO5
Molecular Weight (g/mol) 283.24
MDL Number MFCD01546432
InChI Key CEESSYSSRMOMDE-UHFFFAOYSA-N
Synonym 7-hydroxy-4'-nitroisoflavone, 7-hydroxy-3-4-nitrophenyl-4h-chromen-4-one, 7-hydroxy-3-4-nitrophenyl chromen-4-one, 7-hydroxy-3-4-nitrophenyl-4h-1-benzopyran-4-one, 7-hydroxy-4-nitroisoflavone, 7-hydroxy-4/'-nitroisoflavone, 7-hydroxy-4'-nitro-isoflavone 3c
PubChem CID 7454026
IUPAC Name 7-hydroxy-3-(4-nitrophenyl)chromen-4-one
SMILES OC1=CC=C2C(=O)C(=COC2=C1)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 290°C to 292°C
Color Yellow
Quantity 100 g
Beilstein 385006
Solubility Information Slightly soluble in water.
Formula Weight 283.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 7-Hydroxy-4'-nitroisoflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.