missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-4-(trifluoromethyl)-2H-chromen-2-one, 97%, Thermo Scientific™

Product Code. 10377104
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10377104 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10377104 Supplier Thermo Scientific Maybridge Supplier No. RJF00002DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 575-03-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C10H5F3O3
MDL Number MFCD00037578
Quantity 1g
Synonym 7-hydroxy-4-trifluoromethyl coumarin, 7-hydroxy-4-trifluoromethylcoumarin, 7-hydroxy-4-trifluoromethyl-2h-chromen-2-one, 7-hydroxy-4-trifluoromethyl chromen-2-one, 4-trifluoromethyl umbelliferone, 7-hydroxy-4-trifluoromethyl-chromen-2-one, 2h-1-benzopyran-2-one, 7-hydroxy-4-trifluoromethyl, 7,4-hfc, maybridge1_006885, tfmu
InChI Key CCKWMCUOHJAVOL-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1O)OC(=O)C=C2C(F)(F)F
IUPAC Name 7-hydroxy-4-(trifluoromethyl)chromen-2-one
Molecular Weight (g/mol) 230.142
PubChem CID 5375667
Formula Weight 230.14
Percent Purity 97%
Chemical Name or Material 7-Hydroxy-4-(trifluoromethyl)-2H-chromen-2-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.