missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-4-(trifluoromethyl)coumarin, 98%

Product Code. 11427867
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427867 1 g 1g
11437867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427867 Supplier Thermo Scientific Alfa Aesar Supplier No. A16537.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 575-03-1 | C10H5F3O3 | 230.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 575-03-1
Molecular Formula C10H5F3O3
Molecular Weight (g/mol) 230.142
MDL Number MFCD00037578
InChI Key CCKWMCUOHJAVOL-UHFFFAOYSA-N
Synonym 7-hydroxy-4-trifluoromethyl coumarin, 7-hydroxy-4-trifluoromethylcoumarin, 7-hydroxy-4-trifluoromethyl-2h-chromen-2-one, 7-hydroxy-4-trifluoromethyl chromen-2-one, 4-trifluoromethyl umbelliferone, 7-hydroxy-4-trifluoromethyl-chromen-2-one, 2h-1-benzopyran-2-one, 7-hydroxy-4-trifluoromethyl, 7,4-hfc, maybridge1_006885, tfmu
PubChem CID 5375667
IUPAC Name 7-hydroxy-4-(trifluoromethyl)chromen-2-one
SMILES C1=CC2=C(C=C1O)OC(=O)C=C2C(F)(F)F

Specifications

Melting Point 178°C to 180°C
Quantity 1 g
Formula Weight 230.14
Percent Purity 98%
Chemical Name or Material 7-Hydroxy-4-(trifluoromethyl)coumarin
compliance-icons
Product Identifier
  • 7-Hydroxy-4-(trifluoromethyl)coumarin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Trifluoromethylumbelliferone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.