missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxycoumarin, 98%

Product Code. 11386776
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386776 25 g 25g
11396776 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386776 Supplier Thermo Scientific Alfa Aesar Supplier No. L04082.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-35-6 | C9H6O3 | 162.14 g/mol

7-Hydroxycoumarin is used as an intracellular pH sensitive fluorescent indicator & bloodbrain barrier probe. It is used as a sunscreen agent, and an optical brightener for textiles. It has also been used as a gain medium for dye lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Hydroxycoumarin is used as an intracellular pH sensitive fluorescent indicator & bloodbrain barrier probe. It is used as a sunscreen agent, and an optical brightener for textiles. It has also been used as a gain medium for dye lasers.

Solubility
Soluble in water (slightly at 20°C), 95% ethanol (50 mg/ml), chloroform, acetone, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 93-35-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00006878
InChI Key ORHBXUUXSCNDEV-UHFFFAOYSA-N
Synonym 7-hydroxycoumarin, umbelliferone, 7-hydroxy-2h-chromen-2-one, hydrangin, skimmetin, 7-hydroxycoumarine, 7-oxycoumarin, hydrangine, skimmetine, umbelliferon
PubChem CID 5281426
ChEBI CHEBI:27510
IUPAC Name 7-hydroxychromen-2-one
SMILES OC1=CC=C2C=CC(=O)OC2=C1

Spécification

Melting Point 228°C to 232°C
Quantity 25 g
Beilstein 127683
Merck Index 14,9847
Solubility Information Soluble in water (slightly at 20°C),95% ethanol (50mg/ml),chloroform,acetone,and methanol.
Formula Weight 162.14
Percent Purity 98%
Chemical Name or Material 7-Hydroxycoumarin

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.