missing translation for 'onlineSavingsMsg'
Learn More

7-iodo-2,3-dihydrobenzo[b]furan, 97%, Thermo Scientific™

Product Code. 10112784
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10112784 1 g Amber Glass Bottle 1g
10738815 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10112784 Supplier Thermo Scientific Maybridge Supplier No. CC00911DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 264617-03-0
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD11109311
InChI Key IASDAJCCAFGOCE-UHFFFAOYSA-N
Synonym 7-iodo-2,3-dihydrobenzo b furan, 7-iodo-2,3-dihydrobenzofuran, benzofuran,2,3-dihydro-7-iodo
PubChem CID 33589431
IUPAC Name 7-iodo-2,3-dihydro-1-benzofuran
SMILES C1COC2=C1C=CC=C2I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 246.04
Percent Purity 97%
Chemical Name or Material 7-Iodo-2, 3-dihydrobenzo[b]furan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.