missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxycoumarin, 98+%, ACROS Organics™

Product Code. 10481051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
50g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10481051 50g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10481051 Supplier Thermo Scientific Acros Supplier No. 205120500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 531-59-9
CAS Min % 98.0
CAS Max % 100.0
Melting Point 117°C to 121°C
Color Beige to Pink-Salmon
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C10H8O3
MDL Number MFCD00006876
Quantity 50 g
Beilstein 18, 27
Synonym 7-methoxycoumarin, herniarin, ayapanin, methylumbelliferone, 7-methoxy-2h-chromen-2-one, herniarine, 7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 7-methoxy, coumarin, 7-methoxy, methoxycourmarin, 7
InChI Key LIIALPBMIOVAHH-UHFFFAOYSA-N
SMILES COC1=CC2=C(C=C1)C=CC(=O)O2
IUPAC Name 7-methoxychromen-2-one
Molecular Weight (g/mol) 176.171
PubChem CID 10748
ChEBI CHEBI:5679
Formula Weight 176.17
Percent Purity ≥98.0%
Physical Form Powder
Chemical Name or Material 7-Methoxycoumarin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.