missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxyisoflavone, MedChemExpress

Product Code. 16479186
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479186 Supplier Medchem Express Supplier No. HYN6631

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 7-Methoxyisoflavone is an isoflavone derivative and also an activator of adenosine monophosphate-activated protein kinase (AMPK).

7-Methoxyisoflavone is an isoflavone derivative and also an activator of adenosine monophosphate-activated protein kinase (AMPK).

Specifications

Chemical Name or Material 7-Methoxyisoflavone
CAS 1621-56-3
Color White
Purity Grade Notes Research
Molecular Formula C16H12O3
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (99.10 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C(C2=CC=CC=C2)=COC3=CC(OC)=CC=C13
Molecular Weight (g/mol) 252.26
Formula Weight 252.26
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.