missing translation for 'onlineSavingsMsg'
Learn More

75 cSt Mineral Oil Blank - Trace metals reported, VHG™

Product Code. 16371296
Change view
Click to view available options
Quantity:
500 mL
1 gal
Unit Size:
1 GAL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16371296 500 mL 500mL
17901346 1 gal 1 GAL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16371296 Supplier VHG Labs Supplier No. VHGOIL75500
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

75 cSt Mineral Oil Blank - Trace metals reported
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 75 cSt Mineral Oil Blank
Methods ICP/XRF/RDE
Analyte or Component Names 75 cSt Mineral Oil Blank
Shipping Condition Room Temperature
IUPAC Name 75 cSt Mineral Oil Blank
Solvent or Matrix 75 cSt Hydrocarbon Oil
Quantity 500 mL
compliance-icons
Product Identifier
  • 75 cSt Mineral Oil Blank
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: white mineral oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.