missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody (SPM350) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44730
This item is not returnable.
View return policy
Description
PCNA Monoclonal antibody specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Monkey, S. pombe, Yeast, Zebrafish samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PCNA | |
| Monoclonal | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Chicken | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SPM350 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 0.25 - 0.5 μg/mL | |
| cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
| Rat PCNA/protein A fusion protein (Uniprot: P12004) | |
| 7 mL | |
| Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
| 5111 | |
| Store at 4°C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction