missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-2-naphthol, 94%

Product Code. 10367751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10367751 5 g Glass bottle 5g
10133391 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10367751 Supplier Thermo Scientific Acros Supplier No. 346390050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-46-7 | C10H9NO | 159.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-46-7
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00004031
InChI Key KVHHMYZBFBSVDI-UHFFFAOYSA-N
Synonym 1-amino-7-naphthol, 8-amino-2-naphthol, 2-naphthalenol, 8-amino, 1-amino-7-hydroxynaphthalene, 7-hydroxy-1-naphthylamine, 2-naphthol, 8-amino, 8-amino-2-naphtol, 8-aminonaphth-2-ol, 1-amino-7-naphthol;, 8-amino-2-naphthaleno
PubChem CID 8358
IUPAC Name 8-aminonaphthalen-2-ol
SMILES NC1=C2C=C(O)C=CC2=CC=C1

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point 204.0°C to 207.0°C
Color Brown to Gray
Flash Point >364°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 13, 685
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 159.18
Percent Purity 94%
Physical Form Crystals or Powder
Chemical Name or Material 8-Amino-2-naphthol
Show More Show Less
compliance-icons
Product Identifier
  • 8-Amino-2-naphthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.