missing translation for 'onlineSavingsMsg'
Learn More

8-Anilinonaphthalene-1-sulfonic acid ammonium salt, 98%

Product Code. 15480447
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480447 5 g 5g
15490447 25 g 25g
15400457 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480447 Supplier Thermo Scientific Alfa Aesar Supplier No. J65538.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Fluorescent probe for proteins on polyacrylamide gels | CAS: 28836-03-5 | C16H16N2O3S | 316.38 g/mol

ANS forms an inclusion complex with cyclodextrin. Such model systems are useful to mimic biological recognition and can be studied by measuring the change in fluorescence of free-ANS to complexed-ANS. When ANS enters the hydrophobic core of cyclodestrin, it’s fluorescence increases. It is commonly used to study molecular assemblies of surfactants and amphiphilic polymers because a blue shift of the emission maximum indicates the fluorophore is located in less polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ANS forms an inclusion complex with cyclodextrin. Such model systems are useful to mimic biological recognition and can be studied by measuring the change in fluorescence of free-ANS to complexed-ANS. When ANS enters the hydrophobic core of cyclodestrin, it’s fluorescence increases. It is commonly used to study molecular assemblies of surfactants and amphiphilic polymers because a blue shift of the emission maximum indicates the fluorophore is located in less polar media.

Solubility
Soluble in water, 1N NaOH, and methanol.

Notes
Light & Air Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28836-03-5
Molecular Formula C16H16N2O3S
Molecular Weight (g/mol) 316.38
MDL Number MFCD00012560
InChI Key IPBNQYLKHUNLQE-UHFFFAOYSA-N
Synonym ammonium 8-phenylamino naphthalene-1-sulfonate, 8-anilino-1-naphthalenesulfonic acid ammonium salt, unii-gt7mn8dkd7, gt7mn8dkd7, ammonium 8-anilino-1-naphthalenesulfonate, 1-naphthalenesulfonic acid, 8-phenylamino-, monoammonium salt, 1-naphthalenesulfonic acid, 8-phenylamino-, ammonium salt 1:1, ansa, 1-anilinonaphthalene-8-sulfonic acid ammonium salt, 82-76-8 parent
PubChem CID 120066
IUPAC Name azanium;8-anilinonaphthalene-1-sulfonate
SMILES [NH4+].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1

Specifications

Melting Point 242°C to 244°C
Color Yellow to Green
Quantity 5 g
Beilstein 3581235
Sensitivity Air and light sensitive; Hygroscopic
Solubility Information Soluble in water,1N NaOH,and methanol.
Formula Weight 316.37
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 8-Anilinonaphthalene-1-sulfonic acid ammonium salt
compliance-icons
Product Identifier
  • 8-Anilinonaphthalene-1-sulfonic acid ammonium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 8-Anilinonaphthalene-1-sulfonic acid ammonium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.