missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-1-naphthoic acid, Technical Grade, Maybridge™

Product Code. 10039161
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10039161 1 g Amber Glass Bottle 1g
10796075 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10039161 Supplier Thermo Scientific Maybridge Supplier No. BTB12326DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1729-99-3
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
InChI Key DMEZDDHJCUHENA-UHFFFAOYSA-N
Synonym 8-bromo-1-naphthoic acid, 1-naphthalenecarboxylic acid, 8-bromo, maybridge1_002138, 8-bromo-1-carboxynaphthalene, 8-bromo-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 8-bromo, 8-bromo-naphthalene-1-carboxylic acid
PubChem CID 72873
IUPAC Name 8-bromonaphthalene-1-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)C(=CC=C2)Br

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 251.08
Percent Purity 95%
Chemical Name or Material 8-Bromo-1-naphthoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.