missing translation for 'onlineSavingsMsg'
Learn More

8-(Fmoc-amino)-3,6-dioxaoctanoic Acid, TRC

Product Code. 30403587
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403587 250 mg
30397212 1 g
30394387 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403587 Supplier Toronto Research Chemicals Supplier No. TRCF603515250MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166108-71-0
Molecular Formula C21H23NO6
Molecular Weight (g/mol) 385.41
Synonym Fmoc-AEEA, 12-(9H-Fluoren-9-yl)-10-oxo-3,6,11-trioxa-9-azadodecanoic Acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic Acid, 8-(9-Fluorenylmethoxycarbonylamino)-3,6-dioxaoctanoic Acid, 9-Fluorenylmethoxycarbonyl-8-amino-3,6-dioxaoctanoic Acid
IUPAC Name 2-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]acetic acid
SMILES O=C(NCCOCCOCC(O)=O)OCC1C(C=CC=C2)=C2C3=CC=CC=C13

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H23NO6/c23-20(24)14-27-12-11-26-10-9-22-21(25)28-13-19-17-7-3-1-5-15(17)16-6-2-4-8-18(16)19/h1-8,19H,9-14H2,(H,22,25)(H,23,24)
Formula Weight 385.1525
Chemical Name or Material 8-(Fmoc-amino)-3,6-dioxaoctanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.