missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-DPAT hydrobromide, Tocris Bioscience™

Product Code. 30002179
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30002179

Brand: Tocris Bioscience 0529/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective 5-HT1A agonist; also has moderate affinity for 5-HT7

The standard selective 5-HT1A agonist. Also has moderate affinity for 5-HT7 receptors (pKi is 6.6 at the human 5-HT7 receptor expressed in HEK 293 cells). Reduces hippocampal 5-HT levels following systemic administration in rats in vivo.

R-enantiomer and 7-Hydroxy Isomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76135-31-4
Molecular Formula C16H26BrNO
Molecular Weight (g/mol) 328.29
MDL Number MFCD00055158
InChI Key BATPBOZTBNNDLN-UHFFFAOYNA-N
Synonym 8-hydroxy-dpat hydrobromide, 8-oh-dpat hydrobromide, 8-oh-dpat hrr, 8-hydroxy-2-dipropylaminotetralin hydrobromide, 8-oh-dpat hbr, +/--8-hydroxy-dpat, 7-dipropylamino-5,6,7,8-tetrahydronaphthalen-1-ol hydrobromide, 1-naphthalenol, 7-dipropylamino-5,6,7,8-tetrahydro-, hydrobromide, rac-8-hydroxy-2-dipropylaminotetralin hydrobromide, +/--8-hydroxy-dpat hydrobromide
PubChem CID 6917794
IUPAC Name 7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-1-ol hydrobromide
SMILES Br.CCCN(CCC)C1CCC2=C(C1)C(O)=CC=C2

Specifications

Quantity 50 mg
Formula Weight 328.29
Percent Purity >98%
Chemical Name or Material 8-Hydroxy-DPAT hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.