Learn More
Fisherbrand™ 8 in. Diameter Sieve Receiver
Product Code. 17474003 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (English):
8 in.
Unit Size:
Each
Product Code. Diameter (English) Unit Size
17474003 8 in. Each
1 options
Product Code. 17474003
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use at the bottom of a single sieve or sieve stack
  • Receiving pan for test sieve manufactured in stainless steel. Fits all Fisherbrand 8 in. diameter sieves.
  • Designed to sit under a sieve or sieve stack to catch sample.
Specifications
Voltage 0/1 V
Certifications/Compliance None
Diameter (English) 8 in.