missing translation for 'onlineSavingsMsg'
Learn More

8-oxo-dG Antibody, R&D Systems™

Product Code. 18009351
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18009351 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18009351 Supplier Trevigen Supplier No. 4354MC050
  • ONLINE EXCLUSIVE PRICE
    765.00 €
    723.45 € / 50µL
    Save 41.55 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Mouse Monoclonal Antibody has been used in 38 publications

8-oxo-dG Monoclonal specifically detects 8-oxo-dG in Drosophila∣Human∣Mouse∣Multi-species∣Rat∣Transgenic Mouse samples. It is validated for Bioassay∣ChIP Assay∣Co-immunoprecipitation (Co-IP)∣Comet Assay∣ELISA∣Flow Cytometry∣Immunocytochemistry∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 8-oxo-dG
Applications Bioassay, ChIP Assay, Co-immunoprecipitation (Co-IP), Comet Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 15A3
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:250 dilution, Immunocytochemistry 1:250 dilution
Formulation This antibody is provided as purified immunoglobulin from mouse ascites at 0.5 mg/ml in 1X PBS containing 0.1% sodium azide, 50% glycerol. with 0.1% Sodium Azide
Host Species Mouse
Immunogen 8-oxo-dG-conjugated-KLH
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This mouse monoclonal antibody specifically binds to 8-hydroxy-2'- deoxyguanosine within DNA in H 2 O 2 -treated cells.
Target Species Drosophila, Human, Mouse, Multi-species, Rat, Transgenic Mouse
Content And Storage Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.