missing translation for 'onlineSavingsMsg'
Learn More

8-(p-Toluenesulfonylamino)quinoline, 97%, Thermo Scientific™

Product Code. 15411608
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411608 1 g 1g
15421608 5 g 5g
15431608 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411608 Supplier Thermo Scientific Alfa Aesar Supplier No. H55386.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10304-39-9
Molecular Formula C16H14N2O2S
Molecular Weight (g/mol) 298.36
MDL Number MFCD00168990
InChI Key ZSMKPYXVUIWTCT-UHFFFAOYSA-N
Synonym 8-tosylamino quinoline, 8-p-tosylaminoquinoline, benzenesulfonamide, 4-methyl-n-8-quinolinyl, 8-p-tolylsulfonamido quinoline, 8-p-tolylsulfonylamino quinoline, 8-p-toluenesulfonylamino quinoline, p-toluenesulfonamide, n-8-quinolyl, 4-methyl-n-quinolin-8-yl benzenesulfonamide, 8-4-tolylsulfonylamino quinoline, 4-methyl-n-8-quinolinylbenzenesulfonamide
PubChem CID 82533
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point 154°C to 158°C
Color Brown to White
Quantity 1 g
Beilstein 264159
IUPAC Name 4-methyl-N-(quinolin-8-yl)benzene-1-sulfonamide
Formula Weight 298.36
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 8-(p-Toluenesulfonylamino)quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.