missing translation for 'onlineSavingsMsg'
Learn More
Learn More
High Sulfur EPA Qualification Set: 10x20mL with S @ 100 μg/g, 10x20mL with S @ 500 μg/g, and 20x20mL with S @ 300μg/g in 13 cSt Mineral Oil, VHG™
Click to view available options
Quantity:
40 x 20 mL
Unit Size:
800mL
Description
High Sulfur EPA Qualification Set: 10x20mL with S @ 100 μg/g, 10x20mL with S @ 500 μg/g, and 20x20mL with S @ 300μg/g in 13 cSt Mineral Oil
Specifications
Specifications
| Chemical Name or Material | Reference standards set |
| Methods | XRF |
| Analyte or Component Names | Reference standards set |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| IUPAC Name | Reference standards set |
| Solvent or Matrix | 13 cSt Mineral Oil |
| Quantity | 40 x 20 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction