missing translation for 'onlineSavingsMsg'
Learn More

Tuttnauer 85L Vertical Autoclave

Product Code. 17811097 Shop All Tuttnauer Products
Change view
Click to view available options
Features:
Single Phase, 208 V
Single Phase, 230 V
Three Phase
Three Phase, Cooling
Three Phase, Cooling, Vacuum, Steam Generator, BioHazard Filter (CL3 Waste), Printer
Three Phase, Cooling, Vacuum, Steam Generator, Printer
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Features Unit Size
17811097 Three Phase, Cooling, Vacuum, Steam Generator, BioHazard Filter (CL3 Waste), Printer Each
17861087 Single Phase, 208 V Each
17841077 Single Phase, 230 V Each
17881087 Three Phase Each
17831077 Three Phase, Cooling Each
17801087 Three Phase, Cooling, Vacuum, Steam Generator, Printer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17811097 Supplier Tuttnauer Supplier No. 3870ELVCPVGBHP380D
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The 3870ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.

Features

  • Fo software control
  • PID (Proportional Integral Differential) pressure control
  • Stores the last 200 cycles in built-in memory
  • Two independent flexible PT100 temperature sensors to prevent over-boiling of liquids and explosions of bottles
  • Up to 6 temperature sensors and 4 pressure sensors can be connected
  • 30 Identification Codes and Passwords for access level control
  • The controller and software comply with the 21 CFR part 11 standard

Innovative Multi-Color Display

  • Multi-color display for easier reading
  • Color is used to indicate the stage of the cycle
  • Quick access to important information
  • 26 Languages
  • Built-in view of historical cycle data

Specifications

Capacity (English) 18.7 gal.
Capacity (Metric) 85 L
Depth (Metric) Chamber 70 cm
Diameter (English) Chamber 15.2 in.
Depth (English) Chamber 28 in.
Diameter (Metric) Chamber 38 cm
Length (English) 28 in.
Length (Metric) 70 cm
Product Type Autoclave, Top-loading
Temperature (English) 221°F to 280°F
Temperature (Metric) 105°C to 138°C
Features Three Phase, Cooling, Vacuum, Steam Generator, BioHazard Filter (CL3 Waste), Printer
Material 316L Stainless Steel
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.