missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(4-ethoxyphenyl)-2-chloroanthracene, 98%, Thermo Scientific™

Product Code. 15400118
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
missing translation for 'unitSize'
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15400118

missing translation for 'mfr': Thermo Scientific Alfa Aesar H53369.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135965-21-8
Molecular Formula C30H25ClO2
Molecular Weight (g/mol) 452.98
MDL Number MFCD00190198
InChI Key RFVZBUUPBPFZMH-UHFFFAOYSA-N
Synonym 2-chloro-9,10-bis 4-ethoxyphenyl anthracene, 9,10-bis 4-ethoxyphenyl-2-chloroanthracene, 9,10-bis-4-ethoxyphenyl-2-chloroanthracene
PubChem CID 21715448
IUPAC Name 2-chloro-9,10-bis(4-ethoxyphenyl)anthracene
SMILES CCOC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=C(OCC)C=C2)C2=CC(Cl)=CC=C12

Specifications

Melting Point 264°C to 265°C
Quantity 1 g
Formula Weight 452.98
Percent Purity 98%
Chemical Name or Material 9,10-Bis(4-ethoxyphenyl)-2-chloroanthracene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.