missing translation for 'onlineSavingsMsg'
Learn More

9,10-Diethoxyanthracene, 98%

Product Code. 11492698
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492698 1 g 1g
11402708 5 g 5g
11412708 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492698 Supplier Thermo Scientific Alfa Aesar Supplier No. A16705.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 68818-86-0 | C18H18O2 | 266.34 g/mol

9,10-Diethoxyanthracene acts as a photo sensitizing agent for diaryl iodonium salts. It is also used as a sensitizing dye. It is utilized for the preparation of polymetallic complexes through selective pi-coordination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Diethoxyanthracene acts as a photo sensitizing agent for diaryl iodonium salts. It is also used as a sensitizing dye. It is utilized for the preparation of polymetallic complexes through selective pi-coordination.

Solubility
Soluble in toluene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68818-86-0
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
MDL Number MFCD00209643
InChI Key GJNKQJAJXSUJBO-UHFFFAOYSA-N
Synonym anthracene, 9,10-diethoxy, acmc-1bigo
PubChem CID 111623
IUPAC Name 9,10-diethoxyanthracene
SMILES CCOC1=C2C=CC=CC2=C(C3=CC=CC=C31)OCC

Specifications

Melting Point 149°C to 151°C
Quantity 1 g
Solubility Information Soluble in toluene.
Formula Weight 266.33
Percent Purity 98%
Chemical Name or Material 9,10-Diethoxyanthracene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.