missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dimethylanthracene, 97%

Product Code. 11440627
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440627 5 g 5g
11430627 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440627 Supplier Thermo Scientific Alfa Aesar Supplier No. B22458.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 781-43-1 | C16H14 | 206.29 g/mol

9,10-Dimethylanthracene is used as an antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species. It is also used as chemical rate constant actinometer in singlet molecular oxygen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Dimethylanthracene is used as an antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species. It is also used as chemical rate constant actinometer in singlet molecular oxygen reactions.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 781-43-1
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00001262
InChI Key JTGMTYWYUZDRBK-UHFFFAOYSA-N
Synonym anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
PubChem CID 13076
IUPAC Name 9,10-dimethylanthracene
SMILES CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12

Specifications

Melting Point 182°C to 184°C
Quantity 5 g
Beilstein 1909028
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 206.29
Percent Purity 99%
Chemical Name or Material 9,10-Dimethylanthracene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.