missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%

Product Code. 15492778
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492778 25 g 25g
15402788 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492778 Supplier Thermo Scientific Alfa Aesar Supplier No. H56320.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 286438-45-7 | C37H56Br2 | 660.663 g/mol

It is an intermediate for polymeric light-emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate for polymeric light-emitting diodes.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286438-45-7
Molecular Formula C37H56Br2
Molecular Weight (g/mol) 660.663
MDL Number MFCD03427215
InChI Key KFOUJVGPGBSIFB-UHFFFAOYSA-N
Synonym 2,7-dibromo-9,9-didodecyl-9h-fluorene, 9,9-didodecyl-2,7-dibromofluorene, 9 9-didodecyl-2 7-dibromofluorene, 9h-fluorene, 2,7-dibromo-9,9-didodecyl, 2,7-dibromo-9,9-didodecyl-fluorene, acmc-209h3v, ksc561q2d, 2,7-dibromo-didodecyl-9h-fluorene, 9,9-di-n-dodecyl-2,7-dibromofluorene, 9h-fluorene,2,7-dibromo-9,9-didodecyl
PubChem CID 5140914
IUPAC Name 2,7-dibromo-9,9-didodecylfluorene
SMILES CCCCCCCCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCCCCCCCC

Specifications

Melting Point 50°C to 55°C
Boiling Point 180°C (0.15 mmHg)
Flash Point 110°C (230°F)
Quantity 25 g
Formula Weight 660.65
Percent Purity 98%
Chemical Name or Material 9,9-Di-n-dodecyl-2,7-dibromofluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.