missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-hexyl-2,7-dibromofluorene, 98%

Product Code. 15423508
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423508 100 g 100g
15413508 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423508 Supplier Thermo Scientific Alfa Aesar Supplier No. H56952.22
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 189367-54-2 | C25H32Br2 | 492.339 g/mol

It finds its application as an intermediate for polymeric light-emitting diodes and as an OLED material because of their pure blue emission and efficient electroluminescence coupled with a high charge-carrier mobility and good process ability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an intermediate for polymeric light-emitting diodes and as an OLED material because of their pure blue emission and efficient electroluminescence coupled with a high charge-carrier mobility and good process ability.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189367-54-2
Molecular Formula C25H32Br2
Molecular Weight (g/mol) 492.339
MDL Number MFCD03427215
InChI Key OXFFIMLCSVJMHA-UHFFFAOYSA-N
Synonym 9,9-dihexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene, 2,7-dibromo-9,9-di-n-hexylfluorene, 9h-fluorene, 2,7-dibromo-9,9-dihexyl, 2,7-dibromo-9,9-dihexyl-fluorene, acmc-1c0sw, ksc491i1r, 9,9-di-n-hexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene homopolymer
PubChem CID 3539647
IUPAC Name 2,7-dibromo-9,9-dihexylfluorene
SMILES CCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCC

Specifications

Melting Point 67°C to 71°C
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 492.33
Percent Purity 98%
Chemical Name or Material 9,9-Di-n-hexyl-2,7-dibromofluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.