missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-hexylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15462168
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462168 1 g 1g
15492306 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462168 Supplier Thermo Scientific Alfa Aesar Supplier No. H55817.03
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203927-98-4
Molecular Formula C25H36B2O4
Molecular Weight (g/mol) 422.18
MDL Number MFCD03427279
InChI Key QFWOJNOZWMHNTK-UHFFFAOYSA-N
Synonym 9,9-dihexylfluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid, 7-borono-9,9-dihexyl-fluoren-2-yl boronic acid, boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis, acmc-20alvw, 9,9-dihexyl-fluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diboronic acid, 7-borono-9,9-dihexylfluoren-2-yl boronic acid, 2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluorene
PubChem CID 4524285
SMILES CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O

Specifications

Quantity 1 g
IUPAC Name [7-(dihydroxyboranyl)-9,9-dihexyl-9H-fluoren-2-yl]boronic acid
Formula Weight 422.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 9,9-Di-n-hexylfluorene-2,7-diboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.