missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%

Product Code. p-7034285
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479465 1 g 1g
15489465 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479465 Supplier Thermo Scientific Alfa Aesar Supplier No. H51955.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 196207-58-6 | C41H64B2O4 | 642.579 g/mol

9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester is a reactant in the synthesis of 6,6-(5,5-(9,9-dioctyl-9H-fluorene-2,7-diyl)bis(thiophene-5,2-diyl))bis(2,5-bis(2-ethylhexyl)-3-(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione) (DPP1), a solution-processable nonfullerene electron acceptor used in organic solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester is a reactant in the synthesis of 6,6-(5,5-(9,9-dioctyl-9H-fluorene-2,7-diyl)bis(thiophene-5,2-diyl))bis(2,5-bis(2-ethylhexyl)-3-(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione) (DPP1), a solution-processable nonfullerene electron acceptor used in organic solar cells.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196207-58-6
Molecular Formula C41H64B2O4
Molecular Weight (g/mol) 642.579
MDL Number MFCD16294554
InChI Key FAHIZHKRQQNPLC-UHFFFAOYSA-N
Synonym 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-di-n-octylfluorene, 9,9-di-n-octylfluorene-2,7-diboronic acid bis pinacol ester, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dioctylfluorene, 2-9,9-dioctyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-9,9-dioctyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb260, 9,9-dioctylfluorene-2,7-bis boronic acid pinacol ester, 9,9-dioctylfluorene-2,7-diboronic acid bis pinacol ester, 9,9-dioctyl-9h-fluorene-2,7-diboronic acid bis pinacol ester
PubChem CID 21982074
IUPAC Name 2-[9,9-dioctyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(C3(CCCCCCCC)CCCCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 642.58
Percent Purity 95%
Chemical Name or Material 9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.