missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 9-Aminoacridine

Product Code. 16468756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468756 Supplier MedChemExpress Supplier No. HYB1422

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress 9-Aminoacridine (Aminacrine) is a highly fluorescent dye used as a topical antiseptic and experimentally as a mutagen, an intracellular pH indicator. 9-Aminoacridine is an effective antibacterial agent with caries-disclosing features.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 100 mg
Shipping Condition Room temperature
Solubility DMSO : 27.5 mg/mL (141.58 mM; Need ultrasonic)
Target HBV
Molecular Weight (g/mol) 194.23 g/mol
CAS 90-45-9
Synonym Aminacrine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.