missing translation for 'onlineSavingsMsg'
Learn More

9-Anthracenemethanol, 97%

Product Code. 11492198
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492198 50 g 50g
11482198 10 g 10g
11402208 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492198 Supplier Thermo Scientific Alfa Aesar Supplier No. A15188.18
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1468-95-7 | C15H12O | 208.26 g/mol

An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

Solubility
Soluble in hot methanol very faint turbidity.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1468-95-7
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00001264
InChI Key JCJNNHDZTLRSGN-UHFFFAOYSA-N
Synonym 9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
PubChem CID 73848
IUPAC Name anthracen-9-ylmethanol
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO

Specifications

Melting Point 160°C to 165°C
Quantity 50 g
Beilstein 1873402
Solubility Information Soluble in hot methanol very faint turbidity.
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material 9-Anthracenemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.