missing translation for 'onlineSavingsMsg'
Learn More

9-Anthraldehyde, 98%

Product Code. 11473604
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473604 500 g 500g
11453604 25 g 25g
11463604 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473604 Supplier Thermo Scientific Alfa Aesar Supplier No. A11258.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 642-31-9 | C15H10O | 206.24 g/mol

9-Anthraldehyde is used to prepare asymmetrical tridentate Schiff base ligands, 2-(9-anthrylmethyl-ideneamino)-4-methyl-phenol and functionalized ligand like 2-(anthracen-9-ylidene)-4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione through Knoevenangel condensation with the diphosphine ligand 4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione. It is also used as an intermediate for dyes, pigment, active pharmaceutical ingredients and other organic compounds. Further, it is involved in the Diels Alder reaction with benzenediazonium-2-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Anthraldehyde is used to prepare asymmetrical tridentate Schiff base ligands, 2-(9-anthrylmethyl-ideneamino)-4-methyl-phenol and functionalized ligand like 2-(anthracen-9-ylidene)-4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione through Knoevenangel condensation with the diphosphine ligand 4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione. It is also used as an intermediate for dyes, pigment, active pharmaceutical ingredients and other organic compounds. Further, it is involved in the Diels Alder reaction with benzenediazonium-2-carboxylate.

Solubility
Soluble in toluene.

Notes
Air sensitive. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 642-31-9
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001254
InChI Key YMNKUHIVVMFOFO-UHFFFAOYSA-N
Synonym 9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde
PubChem CID 69504
IUPAC Name anthracene-9-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 102°C to 106°C
Quantity 500 g
Beilstein 639167
Sensitivity Air Sensitive
Solubility Information Soluble in toluene.
Formula Weight 206.24
Percent Purity 98%
Chemical Name or Material 9-Anthraldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.